Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005762.1 | NP_001005762.1 | 9 | 984 | ||
XM_006252241.2 | XP_006252303.1 | 10 | 2668 | |||
XM_006252242.1 | XP_006252304.1 | 9 | 1075 | |||
XM_006252244.2 | XP_006252306.1 | 8 | 1303 | |||
XM_006252246.1 | XP_006252308.1 | 9 | 1020 | |||
XM_008770819.1 | XP_008769041.1 | 7 | 2777 | |||
GenBank mRNA | AF439378.1 | - | 8 | 800 | ||
BC083666.1 | - | 9 | 984 | 7135547 |