Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303457.1 | NP_001290386.1 | 2 | 606 | ||
NM_014657.2 | NP_055472.1 | 3 | 746 | |||
XM_011529114.1 | XP_011527416.1 | 2 | 606 | |||
XM_017028148.1 | XP_016883637.1 | 2 | 606 | |||
GenBank mRNA | AB007866.2 | - | 2 | 695 | ||
AK308164.1 | - | 2 | 594 | |||
BC013121.1 | - | 3 | 709 | 2901089 | ||
BC013755.1 | - | 2 | 559 | 4510329 | ||
BC107778.1 | - | 2 | 603 |