Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006971.1 | NP_001006972.1 | 4, 5 | 377 | ||
XM_006230228.2 | XP_006230290.1 | 5, 6 | 1074 | |||
XM_008759871.1 | XP_008758093.1 | 5, 6 | 1185 | |||
XM_008759872.1 | XP_008758094.1 | 6, 7 | 679 | |||
XM_008759873.1 | XP_008758095.1 | 6, 7 | 754 | |||
XM_008759874.1 | XP_008758096.1 | 6, 7 | 991 | |||
GenBank mRNA | BC083704.1 | - | 4, 5 | 377 | 7190222 |