Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006973.1 | NP_001006974.1 | 2, 3 | 268 | ||
XM_006230739.2 | XP_006230801.1 | 2, 3 | 312 | |||
GenBank mRNA | BC083740.1 | - | 2, 3 | 268 | 7191782 | |
FQ225172.1 | - | 2, 3 | 293 |