Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014663.2 | NP_055478.2 | 22 | 3276 | ||
XM_005271354.3 | XP_005271411.1 | 22 | 3313 | |||
XM_005271355.3 | XP_005271412.1 | 22 | 3231 | |||
XM_005271356.3 | XP_005271413.1 | 23 | 3352 | |||
XM_011542467.2 | XP_011540769.2 | 13 | 2384 | |||
XM_017002912.1 | XP_016858401.1 | 23 | 3421 | |||
GenBank mRNA | AB014577.1 | - | 22 | 3235 | ||
AK294596.1 | - | 8 | 861 | |||
AK300039.1 | - | 14 | 1738 | |||
BC002558.2 | - | 22 | 3244 | 3138875 |