Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006979.1 | NP_001006980.1 | 8 | 1805 | ||
GenBank mRNA | BC083869.1 | - | 8 | 1805 | 7128388 |