Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006981.2 | NP_001006982.2 | 13 | 1087 | ||
XM_006240306.1 | XP_006240368.1 | 11 | 1119 | |||
XM_008764756.1 | XP_008762978.1 | 12 | 1018 | |||
GenBank mRNA | AB504752.1 | - | 6 | 492 | ||
BC083858.1 | - | 13 | 1073 | 7125734 | ||
D90401.1 | - | 13 | 1038 | |||
FQ217447.1 | - | 12 | 1012 | |||
FQ217539.1 | - | 12 | 1012 |