Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxra8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007002.1 | NP_001007003.1 | 5 | 696 | ||
XM_006239574.1 | XP_006239636.1 | 5 | 1064 | |||
XM_006239575.1 | XP_006239637.1 | 5 | 1055 | |||
GenBank mRNA | BC083849.1 | - | 5 | 696 | 7122386 |