Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007006.1 | NP_001007007.1 | 986 | |||
NM_001162408.1 | NP_001155880.1 | 1457 | ||||
XM_008770542.1 | XP_008768764.1 | 6 | 1618 | |||
XM_008770543.1 | XP_008768765.1 | 1816 | ||||
XM_008770544.1 | XP_008768766.1 | 5 | 1414 | |||
GenBank mRNA | BC083646.1 | - | 986 | 7135038 | ||
BC166532.1 | - | 1457 | 7190084 |
Gene Symbol: LOC100909469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751425.3 | XP_003751473.2 | 6 | 1024 | ||
XM_003751426.3 | XP_003751474.1 | 4 | 1816 | |||
XM_008770474.1 | XP_008768696.1 | 6 | 1618 | |||
XM_008770475.1 | XP_008768697.1 | 5 | 1414 |