Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007014.2 | NP_001007015.2 | 2 | 345 | ||
XM_008760164.1 | XP_008758386.1 | 2 | 507 | |||
XM_008760165.1 | XP_008758387.1 | 2 | 496 | |||
XM_008760166.1 | XP_008758388.1 | 2 | 507 | |||
GenBank mRNA | AY303541.1 | - | 2 | 310 | ||
BC086338.1 | - | 309 | ||||
BC086416.1 | - | 2 | 345 | 7130401 |
Gene Symbol: Rbm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170484.1 | NP_001163955.1 | 2 | 325 | ||
XM_008760101.1 | XP_008758323.1 | 2 | 1249 | |||
XM_008760102.1 | XP_008758324.1 | 2 | 1229 | |||
XM_008760103.1 | XP_008758325.1 | 2 | 1086 | |||
XM_008760104.1 | XP_008758326.1 | 2 | 1228 | |||
GenBank mRNA | FM086093.1 | - | 2 | 311 | ||
FQ212306.1 | - | 2 | 431 |