Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007020.2 | NP_001007021.1 | 5 | 957 | ||
NM_001201353.1 | NP_001188282.1 | 6 | 421 | |||
NM_001201354.1 | NP_001188283.1 | 6 | 380 | |||
NM_001201355.1 | NP_001188284.1 | 7 | 521 | |||
XM_006243445.2 | XP_006243507.1 | 6 | 1342 | |||
XM_006243446.2 | XP_006243508.1 | 7 | 1110 | |||
XM_008766200.1 | XP_008764422.1 | 7 | 1013 | |||
GenBank mRNA | BC082036.1 | - | 5 | 912 | 7130894 | |
CB751211.1 | - | 6 | 263 | |||
FM058030.1 | - | 7 | 521 |