Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP3CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000944.4 | NP_000935.1 | 14 | 2603 | ||
NM_001130691.1 | NP_001124163.1 | 13 | 2573 | |||
NM_001130692.1 | NP_001124164.1 | 12 | 2447 | |||
XM_017008365.1 | XP_016863854.1 | 14 | 1907 | |||
GenBank mRNA | AL353950.1 | - | 14 | 2330 | ||
BC025714.1 | - | 13 | 2007 | 5227691 | ||
EU192652.1 | - | 12 | 2447 | |||
EU192653.1 | - | 8 | 1920 | |||
L14778.1 | - | 14 | 2081 |