Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 2 | 319 | ||
NM_001272103.1 | NP_001259032.1 | 2 | 319 | |||
NM_014685.3 | NP_055500.1 | 2 | 319 | |||
GenBank mRNA | AB034989.1 | - | 2 | 262 | ||
AF055001.1 | - | 2 | 259 | |||
AF055003.1 | - | 2 | 62 | |||
AK223233.1 | - | 2 | 262 | |||
AK291565.1 | - | 2 | 262 | |||
AK292088.1 | - | 2 | 262 | |||
AK304800.1 | - | 2 | 262 | |||
BC000086.1 | - | 2 | 254 | 3504515 | ||
BC008320.1 | - | 2 | 253 | 3350209 | ||
BC009739.1 | - | 2 | 262 | 4095997 | ||
BC032673.1 | - | 2 | 261 | 5575914 | ||
BG685245.1 | - | 2 | 250 | |||
CR457116.1 | - | 2 | 164 | |||
D14695.1 | - | 2 | 257 | |||
DA581153.1 | - | 2 | 318 | |||
DQ837586.1 | - | 2 | 261 |