Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007462.1 | NP_001007463.1 | 7 | 911 | ||
GenBank mRNA | BC141316.1 | - | 7 | 910 | 9055928 | |
BC141317.1 | - | 7 | 910 | 9055929 | ||
BC145634.1 | - | 7 | 955 | 9054202 |