Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007525.3 | NP_001007526.3 | 19 | 7418 | ||
NM_001290355.1 | NP_001277284.1 | 21 | 8176 | |||
XM_011527929.1 | XP_011526231.1 | 21 | 7708 | |||
XM_011527930.2 | XP_011526232.1 | 21 | 8194 | |||
XM_011527931.2 | XP_011526233.1 | 20 | 7974 | |||
XM_011527934.1 | XP_011526236.1 | 16 | 5913 | |||
XM_017026669.1 | XP_016882158.1 | 22 | 8261 | |||
GenBank mRNA | BX648940.1 | - | 22 | 8177 |