Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007572.2 | NP_001007573.1 | 11 | 1383 | ||
GenBank mRNA | AK085479.1 | - | 11 | 1429 | ||
BC110554.1 | - | 11 | 1383 | 40047811 | ||
BC110555.1 | - | 11 | 1383 | 40047816 |