Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007606.1 | NP_001007607.1 | 8, 9 | 1165 | ||
XM_006233136.2 | XP_006233198.1 | 8, 9 | 1200 | |||
GenBank mRNA | BC061765.1 | - | 8, 9 | 1165 | 5620893 | |
FQ233851.1 | - | 1, 2 | 150 |