Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007617.1 | NP_001007618.1 | 3 | 495 | ||
XM_008769444.1 | XP_008767666.1 | 3 | 1196 | |||
GenBank mRNA | BC081899.1 | - | 3 | 495 | 7110518 |