Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000952.4 | NP_000943.1 | 2 | 1492 | ||
NM_001164721.1 | NP_001158193.1 | 3 | 1436 | |||
NM_001164722.2 | NP_001158194.1 | 2 | 1354 | |||
NM_001164723.2 | NP_001158195.1 | 3 | 1563 | |||
GenBank mRNA | AK290240.1 | - | 2 | 1238 | ||
AK292053.1 | - | 3 | 1436 | |||
BC013816.2 | - | 1 | 1141 | 4343214 | ||
BC063000.1 | - | 2 | 1228 | 5176867 | ||
CD013879.1 | - | 3 | 1292 | |||
M80436.1 | - | 2 | 1367 |
Set Membership: |
G-protein coupled receptor |