Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007621.1 | NP_001007622.1 | 3 | 590 | ||
XM_006251946.2 | XP_006252008.1 | 3 | 455 | |||
XM_006251947.2 | XP_006252009.1 | 4 | 523 | |||
XM_006251948.2 | XP_006252010.1 | 4 | 501 | |||
GenBank mRNA | BC079263.1 | - | 3 | 590 | 7133513 | |
FQ211258.1 | - | 3 | 514 |