Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142685.1 | NP_001136157.1 | 12 | 1169 | ||
NM_014715.3 | NP_055530.2 | 3 | 501 | |||
XM_005271736.4 | XP_005271793.2 | 13 | 2199 | |||
XM_011543072.2 | XP_011541374.1 | 13 | 1401 | |||
XM_011543073.2 | XP_011541375.2 | 13 | 1137 | |||
XM_011543075.2 | XP_011541377.1 | 4 | 347 | |||
XM_017018595.1 | XP_016874084.1 | 13 | 1118 | |||
XM_017018596.1 | XP_016874085.1 | 11 | 1395 | |||
XM_017018597.1 | XP_016874086.1 | 13 | 2208 | |||
GenBank mRNA | AB018255.2 | - | 3 | 422 | ||
AB079856.1 | - | 3 | 501 | |||
AB088416.1 | - | 3 | 422 | |||
AL833062.2 | - | 11 | 971 | |||
AY194287.1 | - | 3 | 425 | |||
BC000277.1 | - | 5 | 415 | |||
BC051236.1 | - | 7 | 625 | |||
BC065371.1 | - | 7 | 625 | |||
BC104898.1 | - | 2 | 156 | 8143901 | ||
BC113429.1 | - | 2 | 156 | 8322481 | ||
EF127492.1 | - | 12 | 1169 |