Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007634.1 | NP_001007635.1 | 2 | 1445 | ||
GenBank mRNA | BC083637.1 | - | 2 | 1445 | 7134766 |