Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007640.1 | NP_001007641.1 | 3 | 364 | ||
XM_006233320.2 | XP_006233382.1 | 4 | 1494 | |||
XM_006233321.2 | XP_006233383.1 | 4 | 480 | |||
GenBank mRNA | BC079404.1 | - | 3 | 364 | 7112817 | |
FQ212179.1 | - | 4 | 455 |