Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2j2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007655.1 | NP_001007656.1 | 3 | 543 | ||
XM_006239554.2 | XP_006239616.1 | 3 | 395 | |||
XM_006239555.2 | XP_006239617.1 | 4 | 730 | |||
XM_006239557.2 | XP_006239619.1 | 2 | 390 | |||
XM_006239558.2 | XP_006239620.1 | 2 | 232 | |||
XM_008764352.1 | XP_008762574.1 | 3 | 553 | |||
XM_008764353.1 | XP_008762575.1 | 3 | 1623 | |||
XM_008764354.1 | XP_008762576.1 | 4 | 734 | |||
XM_008764355.1 | XP_008762577.1 | 4 | 1679 | |||
GenBank mRNA | BC079115.1 | - | 3 | 543 | 7126201 |