Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303627.1 | NP_001290556.1 | 2 | 180 | ||
NM_001303628.1 | NP_001290557.1 | 2 | 463 | |||
NM_014730.3 | NP_055545.1 | 2 | 463 | |||
XM_011539031.1 | XP_011537333.1 | 2 | 4878 | |||
XM_011539032.1 | XP_011537334.1 | 3 | 4566 | |||
GenBank mRNA | BC000371.2 | - | 2 | 412 | 2819615 | |
BC016297.2 | - | 2 | 414 | 3832682 | ||
BG718506.1 | - | 2 | 468 | |||
BI458950.1 | - | 2 | 182 | |||
BU845446.1 | - | 441 | ||||
D63486.1 | - | 2 | 418 |