Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007667.2 | NP_001007668.1 | 6 | 842 | ||
XM_008773228.1 | XP_008771450.1 | 6 | 1842 | |||
XM_008773229.1 | XP_008771451.1 | 6 | 1238 | |||
XM_008773230.1 | XP_008771452.1 | 7 | 877 | |||
GenBank mRNA | BC060588.1 | - | 6 | 844 | 6921572 | |
FQ222039.1 | - | 6 | 724 | |||
FQ223333.1 | - | 1 | 118 | |||
FQ229984.1 | - | 6 | 726 |