Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007671.1 | NP_001007672.1 | 2 | 481 | ||
XM_006246730.2 | XP_006246792.1 | 2 | 259 | |||
GenBank mRNA | BC079177.1 | - | 2 | 481 | 7130324 |