Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007672.1 | NP_001007673.1 | 5, 6 | 536 | ||
GenBank mRNA | BC078932.1 | - | 5, 6 | 536 | 7107480 | |
FQ214419.1 | - | 5, 6 | 690 |