Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 9 | 861 | ||
NM_170774.1 | NP_739580.1 | 8 | 719 | |||
XM_005260895.3 | XP_005260952.1 | 9 | 927 | |||
XM_011529410.1 | XP_011527712.1 | 9 | 861 | |||
XM_011529411.1 | XP_011527713.1 | 9 | 933 | |||
XM_011529412.2 | XP_011527714.1 | 8 | 782 | |||
XM_017028149.1 | XP_016883638.1 | 8 | 756 | |||
XM_017028150.1 | XP_016883639.1 | 10 | 1064 | |||
XM_017028151.1 | XP_016883640.1 | 10 | 1031 | |||
XM_017028152.1 | XP_016883641.1 | 10 | 1108 | |||
XM_017028153.1 | XP_016883642.1 | 8 | 809 | |||
GenBank mRNA | AK222650.1 | - | 9 | 824 | ||
AK291458.1 | - | 9 | 816 | |||
AY154470.1 | - | 9 | 861 | |||
AY154471.1 | - | 6 | 688 | |||
AY154472.1 | - | 8 | 719 | |||
BC110385.1 | - | 6 | 688 | |||
BC117118.1 | - | 7 | 698 | 40125669 | ||
BC117120.1 | - | 7 | 698 | 40125671 | ||
CR627436.1 | - | 8 | 4068 | |||
D79990.1 | - | 9 | 861 |