Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciapin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007689.1 | NP_001007690.1 | 6 | 910 | ||
XM_006255106.2 | XP_006255168.1 | 6 | 997 | |||
XM_006255107.2 | XP_006255169.1 | 6 | 709 | |||
XM_006255108.2 | XP_006255170.1 | 6 | 712 | |||
GenBank mRNA | BC083753.1 | - | 6 | 910 | 7192604 | |
FQ211546.1 | - | 6 | 694 | |||
FQ230885.1 | - | 6 | 922 |