Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099.4 | NP_001090.2 | 10 | 1576 | ||
NM_001292037.1 | NP_001278966.1 | 9 | 1477 | |||
GenBank mRNA | BC008493.1 | - | 10 | 1533 | 4272289 | |
BC016344.1 | - | 10 | 1540 | 3950907 | ||
M24902.1 | - | 10 | 1510 | |||
M34840.1 | - | 10 | 1501 | |||
X52174.1 | - | 10 | 1510 | |||
X53605.1 | - | 10 | 1523 |