Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007693.1 | NP_001007694.1 | 4 | 731 | ||
XM_006230984.2 | XP_006231046.1 | 4 | 960 | |||
XM_006230985.2 | XP_006231047.1 | 5 | 876 | |||
XM_006230986.2 | XP_006231048.1 | 4 | 780 | |||
XM_006230987.2 | XP_006231049.1 | 4 | 715 | |||
XM_006230988.2 | XP_006231050.1 | 5 | 747 | |||
GenBank mRNA | BC062803.1 | - | 4 | 731 | 6890943 | |
FQ211550.1 | - | 4 | 740 |