Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007693.1 | NP_001007694.1 | 4 | 1214 | ||
XM_006230984.2 | XP_006231046.1 | 4 | 1443 | |||
XM_006230985.2 | XP_006231047.1 | 5 | 1359 | |||
XM_006230986.2 | XP_006231048.1 | 4 | 1263 | |||
XM_006230987.2 | XP_006231049.1 | 4 | 1198 | |||
XM_006230988.2 | XP_006231050.1 | 5 | 1230 | |||
GenBank mRNA | BC062803.1 | - | 4 | 1214 | 6890943 | |
FQ211550.1 | - | 4 | 1223 |