Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam203a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007707.1 | NP_001007708.1 | 6 | 1936 | ||
GenBank mRNA | BC079158.1 | - | 6 | 1936 | 7129210 |