Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ribc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007715.1 | NP_001007716.1 | 7, 8 | 1224 | ||
XM_006256799.2 | XP_006256861.1 | 6, 7 | 1411 | |||
XM_006256800.2 | XP_006256862.1 | 7, 8 | 1364 | |||
XM_006256801.2 | XP_006256863.1 | 7, 8 | 1375 | |||
XM_006256802.2 | XP_006256864.1 | 7, 8 | 1279 | |||
XM_006256803.2 | XP_006256865.1 | 7, 8 | 1282 | |||
XM_006256804.2 | XP_006256866.1 | 7, 8 | 1264 | |||
XM_006256805.2 | XP_006256867.1 | 7, 8 | 1260 | |||
GenBank mRNA | BC078998.1 | - | 7, 8 | 1224 | 7112763 |