Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007731.1 | NP_001007732.1 | 5 | 1162 | ||
XM_006253320.2 | XP_006253382.1 | 5 | 1215 | |||
GenBank mRNA | BC078959.1 | - | 5 | 1162 | 7110849 |