Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007752.1 | NP_001007753.1 | 5 | 652 | ||
GenBank mRNA | BC079390.1 | - | 5 | 652 | 7111791 |