Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007757.1 | NP_001007758.1 | 3 | 1009 | ||
XM_006257228.2 | XP_006257290.1 | 4 | 1130 | |||
XM_008773423.1 | XP_008771645.1 | 7 | 2425 | |||
XM_008773424.1 | XP_008771646.1 | 7 | 3427 | |||
XM_008773425.1 | XP_008771647.1 | 4 | 1154 | |||
GenBank mRNA | BC081893.1 | - | 3 | 1009 | 7109901 |