Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007758.1 | NP_001007759.1 | 8 | 1068 | ||
GenBank mRNA | BC082111.1 | - | 8 | 1068 | 7135272 |