Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310135.1 | NP_001297064.1 | 33 | 6451 | ||
XM_011537430.2 | XP_011535732.1 | 29 | 4283 | |||
XM_011537431.2 | XP_011535733.1 | 27 | 3940 | |||
XM_017021254.1 | XP_016876743.1 | 32 | 6301 | |||
XM_017021255.1 | XP_016876744.1 | 31 | 6166 | |||
XM_017021256.1 | XP_016876745.1 | 32 | 6565 | |||
GenBank mRNA | BC014342.2 | - | 6 | 858 | ||
BC103914.2 | - | 15 | 2021 | 40006180 | ||
BC103915.1 | - | 14 | 1800 | 40006182 | ||
BC103916.2 | - | 14 | 1800 | 40006183 | ||
BX161415.1 | - | 13 | 1735 |