Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007796.1 | NP_001007797.1 | 2 | 359 | ||
GenBank mRNA | BC085781.1 | - | 2 | 402 | 7107433 | |
DQ233644.1 | - | 2 | 416 |