Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007798.1 | NP_001007799.1 | 3 | 336 | ||
GenBank mRNA | AY787790.1 | - | 3 | 336 | ||
FQ215667.1 | - | 479 |