Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003466.3 | NP_003457.1 | 10, 11 | 1363 | ||
NM_013952.3 | NP_039246.1 | 10, 11 | 1284 | |||
NM_013953.3 | NP_039247.1 | 8, 9 | 1053 | |||
NM_013992.3 | NP_054698.1 | 7, 8 | 951 | |||
XM_011511790.1 | XP_011510092.1 | 10, 11 | 1395 | |||
XM_011511792.1 | XP_011510094.1 | 10, 11 | 1316 | |||
XM_011511793.1 | XP_011510095.1 | 9, 10 | 1206 | |||
XM_011511794.1 | XP_011510096.1 | 9, 10 | 1175 | |||
GenBank mRNA | AK292191.1 | - | 10, 11 | 1360 | ||
AK307173.1 | - | 2, 3 | 201 | |||
BC001060.2 | - | 10, 11 | 1354 | 2963877 | ||
CR936798.1 | - | 1 | 5366 | |||
KC733810.1 | - | 10, 11 | 1460 | |||
KJ545852.1 | - | 1 | 206 | |||
KJ545853.1 | - | 1 | 206 | |||
KJ545855.1 | - | 1 | 206 | |||
KJ545856.1 | - | 1 | 205 | |||
KJ545857.1 | - | 1 | 206 | |||
KJ545858.1 | - | 1 | 206 | |||
KJ545859.1 | - | 1 | 206 | |||
KJ545860.1 | - | 1 | 209 | |||
KJ545861.1 | - | 1 | 230 | |||
KJ545862.1 | - | 1 | 206 | |||
KJ545863.1 | - | 1 | 206 | |||
KJ545864.1 | - | 1 | 222 | |||
KJ545867.1 | - | 1 | 207 | |||
KJ545868.1 | - | 1 | 206 | |||
KJ545869.1 | - | 1 | 224 | |||
KJ545870.1 | - | 1 | 206 | |||
KJ545871.1 | - | 1 | 206 | |||
KJ545873.1 | - | 1 | 205 | |||
KJ545875.1 | - | 1 | 224 | |||
KJ545883.1 | - | 1 | 206 | |||
KJ545884.1 | - | 1 | 206 | |||
KU178404.1 | - | 9, 10 | 1197 | |||
KU178405.1 | - | 8, 9 | 1008 | |||
KU178406.1 | - | 8, 9 | 999 | |||
L19606.1 | - | 9, 10 | 1207 | |||
S77904.1 | - | 10, 11 | 1278 | |||
S77905.1 | - | 8, 9 | 1047 | |||
S77906.2 | - | 7, 8 | 945 | |||
X69699.1 | - | 10, 11 | 1357 |