Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cir1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007799.1 | NP_001007800.1 | 10 | 1142 | ||
GenBank mRNA | BC085869.1 | - | 10 | 1212 | 7191148 |