Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008231.2 | NP_001008232.2 | 25 | 3310 | ||
GenBank mRNA | AK009382.1 | - | 10 | 1132 | ||
AK122266.1 | - | 23 | 6228 | |||
AY426536.1 | - | 25 | 3339 | |||
BC037720.1 | - | 9 | 1031 |