Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008285.1 | NP_001008286.1 | 8 | 655 | ||
XM_006248331.2 | XP_006248393.1 | 9 | 869 | |||
XM_006248332.2 | XP_006248394.1 | 8 | 823 | |||
XM_006248334.1 | XP_006248396.1 | 6 | 552 | |||
XM_008768719.1 | XP_008766941.1 | 8 | 753 | |||
XM_008768721.1 | XP_008766943.1 | 6 | 1084 | |||
GenBank mRNA | BC085954.1 | - | 8 | 655 | 7134530 | |
FQ210155.1 | - | 8 | 721 |