Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314021.1 | NP_001300950.1 | 3 | 382 | ||
NM_014777.3 | NP_055592.2 | 3 | 351 | |||
XM_005273360.3 | XP_005273417.1 | 3 | 320 | |||
GenBank mRNA | AK291013.1 | - | 3 | 351 | ||
BC059381.1 | - | 3 | 326 | |||
BC114559.1 | - | 3 | 304 | 40082897 | ||
BC114963.1 | - | 3 | 273 | 40082900 | ||
D50923.2 | - | 3 | 333 | |||
DC369323.1 | - | 3 | 351 |