Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008309.1 | NP_001008310.1 | 6 | 785 | ||
XM_006231835.2 | XP_006231897.1 | 6 | 1055 | |||
GenBank mRNA | BC085343.1 | - | 6 | 785 | 7303981 | |
FQ220452.1 | - | 6 | 789 | |||
FQ229173.1 | - | 6 | 791 |