Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014783.4 | NP_055598.1 | 1 | 828 | ||
NM_199357.1 | NP_955389.1 | 1 | 828 | |||
GenBank mRNA | BC039563.2 | - | 1 | 683 | 3913511 | |
BC050545.1 | - | 1 | 783 | |||
BC063444.1 | - | 1 | 779 | |||
D87717.2 | - | 1 | 828 |
Gene Symbol: ARHGAP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039841.1 | NP_001034930.1 | 1 | 251 | ||
GenBank mRNA | BC105788.1 | - | 1 | 251 | 4072723 |