Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008322.1 | NP_001008323.1 | 4 | 533 | ||
XM_006241453.2 | XP_006241515.1 | 4 | 559 | |||
GenBank mRNA | BC085331.1 | - | 4 | 533 | 7106911 |